Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cask Mouse anti-Mouse, Rat, ATTO 488, Clone: PT55, Abnova™

Catalog No. 89338913 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-913 Supplier Abnova Corporation Supplier No. MAB19409

Mouse Monoclonal Antibody.

Cask Monoclonal antibody specifically detects Cask in Mouse, Rat samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen Cask
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PT55
Conjugate Atto 488
Description calcium/calmodulin-dependent serine protein kinase (MAGUK family)
Gene Cask
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 318-415 of rat Cask
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 29647
Target Species Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.