Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASKIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563228 Supplier Invitrogen™ Supplier No. PA563228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKEQELVQAV KAEDVGTAQR LLQRPRPGKA KLLGSTKKIN VNFQDPDGFS A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Calcium/calmodulin-dependent serine protein kinase (CASK) is a conserved multi-domain scaffolding protein involved in brain development, synapse formation, and establishment of cell polarity. CASKINs (CASK interacting proteins) interact with CASK and is thought to play a role in CASK function, specifically by coupling CASK to distinct downstream effectors. CASKIN1 is a multidomain protein containing six N-terminal ankyrin repeats, one SH3 domain, and two sterile alpha motif domains followed by a long proline-rich sequence and a short conserved C-terminal domain. CASKIN1 coassembles with CASK on the cytoplasmic tail of neurexin 1, suggesting that CASKIN and CASK coat the cytoplasmic tails of neurexins and other cell-surface proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASKIN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CASKIN1
Gene Accession No. Q8WXD9
Gene Alias ANKS5A; CASK interacting protein 1; CASKIN1; Caskin-1; CASK-interacting protein 1; KIAA1306
Gene Symbols CASKIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CASKIN1. Recombinant protein control fragment (Product #RP-101673).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57524
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.