Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASKIN2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152629IR
Description
CASKIN2 Polyclonal antibody specifically detects CASKIN2 in Human samples. It is validated for Western BlotSpecifications
CASKIN2 | |
Polyclonal | |
Western Blot | |
ANKS5B, CASK interacting protein 2, caskin-2, FLJ21609, KIAA1139 | |
The immunogen for this product maps to a region between residue 1100 to 1150 of human CASK Interacting Protein 2 using the numbering given in entry NP_065804.2 (GeneID 57513). | |
0.1 mL | |
Signal Transduction | |
57513 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction