Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASKIN2 Monoclonal Antibody (OTI7C5), TrueMAB™, OriGene
SDP

Catalog No. 502543758
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3758 Supplier OriGene Supplier No. CF812219
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CASKIN2 Monoclonal antibody specifically detects CASKIN2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen CASKIN2
Applications Western Blot
Classification Monoclonal
Clone OTI7C5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MYO18A
Gene Accession No. Q92614, Q9JMH9
Gene Alias 1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 600-1000 of human MYO18A (NP_976063) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360013, 360570, 399687
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.