Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CASKIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572753
Description
CASKIN2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This gene encodes a large protein that contains six ankyrin repeats, as well as a Src homology 3 (SH3) domain and two sterile alpha motif (SAM) domains, which may be involved in protein-protein interactions. The C-terminal portion of this protein is proline-rich and contains a conserved region. A related protein interacts with calcium/calmodulin-dependent serine protein kinase (CASK). Alternative splicing results in multiple transcript variants.
Specifications
CASKIN2 | |
Polyclonal | |
Unconjugated | |
CASKIN2 | |
1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57513 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8WXE0 | |
CASKIN2 | |
CASKIN2 antibody for a 19 amino acid synthetic peptide near the amino terminus of human CASKIN2. The immunogen is within amino acids 380 - 430 of CASKIN2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction