Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASKIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584882 Supplier Invitrogen™ Supplier No. PA584882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APWAFSYLAG PPATPPDPPR PKRRSHSLSR PGPTEGDAEG EAEGPVGSTL GSYATLTRRP GRSALVRTSP SVTPTPARGT PRSQSFALR Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.

This gene encodes a large protein that contains six ankyrin repeats, as well as a Src homology 3 (SH3) domain and two sterile alpha motif (SAM) domains, which may be involved in protein-protein interactions. The C-terminal portion of this protein is proline-rich and contains a conserved region. A related protein interacts with calcium/calmodulin-dependent serine protein kinase (CASK). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASKIN2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CASKIN2
Gene Accession No. Q8WXE0
Gene Alias 1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139
Gene Symbols CASKIN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CASKIN2. Recombinant protein control fragment (Product #RP-108255).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.