Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CASP3 Mouse anti-Human, Mouse, Rat, Clone:3D4D10, Abnova™
Mouse monoclonal antibody raised against recombinant human CASP3.
Supplier: Abnova Corporation MAB16670
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 6, 7 and 9, and the protein itself is processed by caspases 8, 9 and 10. It is the predominant caspase involved in the cleavage of amyloid-beta 4A precursor protein, which is associated with neuronal death in Alzheimer′s disease. Alternative splicing of this gene results in two transcript variants that encode the same protein. [provided by RefSeq]Specifications
CASP3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
CASP3 | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
3D4D10 | |
caspase 3, apoptosis-related cysteine peptidase | |
In PBS (0.05% sodium azide) | |
CPP32, CPP32B, SCA-1 | |
Recombinant protein corresponding to amino acid 29-175 of human CASP3 from E. coli. | |
836 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction