Learn More
CASP4 Mouse anti-Human, Clone: OTI4A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805590

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.Specifications
CASP4 | |
Monoclonal | |
Unconjugated | |
CASP4 | |
ICE(rel)II, ICEREL-II, ICH-2, Mih1, Mih1/TX, TX | |
Mouse | |
Affinity Chromatography | |
RUO | |
837 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P49662 | |
CASP4 | |
Human recombit protein fragment corresponding to amino acids 81-270 of human CASP4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.