Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP8, Mouse anti-Human, Clone: C15, Abnova™

Catalog No. 89998859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-859 Supplier Abnova Corporation Supplier No. MAB16115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen CASP8
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C15
Conjugate Unconjugated
Dilution Immunoprecipitation Immunocytochemistry Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS (10% glycerol, 0.02% sodium azide).
Gene caspase 8, apoptosis-related cysteine peptidase
Gene Alias ALPS2B/CAP4/FLICE/FLJ17672/MACH/MCH5/MGC78473
Gene Symbols CASP8
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 181-478 of human CASP8.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 841
Test Specificity Recognizes the p18 subunit of human CASP8.
Target Species Human
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.