Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP215713
Description
Caspase-1 Polyclonal specifically detects Caspase-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Caspase-1 | |
Polyclonal | |
Unconjugated | |
PBS (pH7), 20% Glycerol with 0.025% Proclin 300 | |
CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 | |
Rabbit | |
45 kDa | |
0.1 mL | |
Apoptosis, Cancer, Caspases | |
834 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.64mg/mL | |
Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P29466 | |
CASP1 | |
Recombinant protein encompassing a sequence within the center region of human Caspase 1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction