Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$192.47 - $517.59
Specifications
| Antigen | Caspase-1 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056564T
![]() |
Novus Biologicals
NB10056564SS |
0.025 mg |
Each for $192.47
|
|
|||||
NB10056564
![]() |
Novus Biologicals
NB10056564 |
0.1 mg |
Each for $517.59
|
|
|||||
Description
Caspase-1 Polyclonal specifically detects Caspase-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Caspase-1 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P29466 | |
| 834 | |
| A synthetic peptide corresponding to amino acids 371-390 RKVRFSFEQPDGRAQMPTTE of human Caspase-1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Caspases | |
| PBS with 0.05% Sodium Azide | |
| CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 | |
| CASP1 | |
| IgG | |
| Protein G purified | |
| The antibody recognizes full-length Caspase-1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title