Learn More
Caspase 1/p20/p10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Caspase 1/p20/p10 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3966
![]() ![]() |
Proteintech Group Inc
229151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
22915-1-AP can recognize all five isoforms of caspase 1. It also recognizes p45 procaspase and the mature fragments p10 and p20.
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.Specifications
Caspase 1/p20/p10 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P29452, P29466, P43527 | |
12362, 25166, 834 | |
Caspase 1/p20/p10 Fusion Protein Ag11143 | |
Primary | |
-20°C | |
CASP1 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CASP 1, CASP1, Caspase 1, Caspase 1/p20/p10, Caspase1, ICE, IL 1 beta converting enzyme, IL 1BC, IL1BC, IL1BCE, Interleukin 1 beta convertase, p10, p20, P45 | |
Casp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.