Learn More
Invitrogen™ Caspase 1 Polyclonal Antibody
Description
Caspase 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | Caspase 1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | CASP1 |
| Gene Accession No. | P29452, P29466, Q9N2I1 |
| Gene Alias | CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45 |
| Gene Symbols | CASP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.