Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 1 Polyclonal Antibody

Catalog No. PIPA5122284
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122284 20 μL
PIPA5119001 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122284 Supplier Invitrogen™ Supplier No. PA5122284
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CASP1
Gene Accession No. P29466, Q9N2I1
Gene Alias CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45
Gene Symbols CASP1
Host Species Rabbit
Immunogen Recombinant protein Caspase 1. The antigen corresponds to amino acid range 120-297 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397319, 834
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.