Learn More
Invitrogen™ Caspase 10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120032
Description
Positive test controls include: Jurkat, NIH/3T3. Immunogen sequence: VKTFLEALPQ ESWQNKHAGS NGNRATNGAP SLVSRGMQGA SANTLNSETS TKRAAVYRMN RNHRGLCVIV NNHSFTSLKD RQGTHKDAEI LSHVFQWLGF TVHIHNNVTK VEMEMVLQKQ KCNPAHADGD CFVFCILTHG RFGAVYSSDE ALIPIREIMS HFTALQCPRL AEKPKLFFIQ ACQGEEIQPS VSIEADALNP EQAPTSLQDS IPAEADFLLG LATVPGYVSF RHVEEGSWYI QSLCNHLKKL VPRHEDILSI LTAVNDDVSR RVDKQGTKKQ MPQPAFTLRK KLVFPVPLDA LS.
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 3 and 7, and the protein itself is processed by caspase 8. Mutations in this gene are associated with apoptosis defects seen in type II autoimmune lymphoproliferative syndrome. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| Caspase 10 | |
| Polyclonal | |
| Unconjugated | |
| CASP10 | |
| ALPS2; Apoptotic protease Mch-4; CASP10; CASP-10; caspase 10; caspase 10 apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine protease; caspase-10; Caspase-10 subunit p12; Caspase-10 subunit p23/17; FADD-like ICE2; FAS-associated death domain protein interleukin-1B-converting enzyme 2; FLICE2; ICE-like apoptotic protease 4; interleukin-1B-converting enzyme 2; MCH4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 843 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92851 | |
| CASP10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-521 of human Caspase-10 (NP_1167592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.