Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-11 Antibody (17D9) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody has been used in 47 publications

$244.50 - $562.50

Specifications

Antigen Caspase-11
Clone 17D9
Concentration 1.0 mg/mL
Dilution Western Blot 5 ug/ml, Flow Cytometry reported in scientific literature (PMID 27708283), ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 10791975), Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500, Immunoblotting reported in scientific literature (PMID 28345580), Microarray, Knockout Validated, KnockDown Validated
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB12010454T
SDP
View Documents
Novus Biologicals
NB12010454SS
0.025 mL
Each for $244.50
Only null left
Add to Cart
 
NB12010454
SDP
View Documents
Novus Biologicals
NB12010454
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

Caspase-11 Monoclonal specifically detects Caspase-11 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Microarray, Knockout Validated, Knockdown Validated.
Specifications

Specifications

Caspase-11
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown
Unconjugated
Rat
Human, Mouse, Rat, Bovine
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl
Casp4
IgG2a
Protein G purified
35-43 kDa
17D9
Western Blot 5 ug/ml, Flow Cytometry reported in scientific literature (PMID 27708283), ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 10791975), Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500, Immunoblotting reported in scientific literature (PMID 28345580), Microarray, Knockout Validated, KnockDown Validated
Monoclonal
Purified
RUO
P70343
12363
A recombinant p30 subunit of mouse Caspase 11. [UniProt# P70343]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.