Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-11 Antibody (17D9), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB12010454MFV450
Description
Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Human, Mouse, Rat, Bovine samples. It is validated for Flow CytometrySpecifications
Caspase-11 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
12363 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
17D9 | |
Flow Cytometry | |
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
A recombinant p30 subunit of mouse Caspase 11. [UniProt# P70343] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction