Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-11 Antibody (4E11), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280100FR
This item is not returnable.
View return policy
Description
Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
Caspase-11 | |
Monoclonal | |
DyLight 680 | |
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
Recombinant mouse caspase-11 (p20 fragment). | |
0.1 mL | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
400000000000 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
12363 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction