Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-11 Antibody (8A5) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280099
Description
Caspase-11 Monoclonal specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.Specifications
Caspase-11 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
Rat | |
Protein G purified | |
RUO | |
12363 | |
Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
8A5 | |
Unconjugated | |
PBS | |
Casp4 | |
Recombinant mouse Caspase-11 (p20 fragment). | |
100 μg | |
Primary | |
Recognizes mouse and rat full-length Caspase-11 protein. Detects two bands of ∽38kDa and ∽43kDa by Western blot. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction