Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 11 Monoclonal Antibody (17D9), eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

$346.50

Specifications

Antigen Caspase 11
Clone 17D9
Concentration 0.5 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-138-95
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
14993582
100 μg
$462.00
$346.50 / Each of 1

Save $115.50

25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
Description

Description

Description: The monoclonal antibody 17D9 recognizes mouse Caspase-11, a cysteine protease that plays a role in apoptosis and inflammation. Caspase-11 exists as a 43 kDa and 38 kDa pro-caspase form and a 30 and 10 kDa active form. The 17D9 antibody recognizes p43, p38 and p30 forms. Caspase-11 is an upstream protease that activates other caspases, such as caspase-3, -7 and -1, which in turn activate IL-1beta. Under Caspase-11-deficient conditions, cells are unable to process or secrete IL-1beta in response to stimuli and show defects in motility, which are due to disruption of its interaction with Aip1, an actin binding protein. Caspase-11 is homologous to human Caspase-4 (around 60%). Unlike other caspases which tend to be constitutively expressed, caspase-11 is upregulated by LPS treatment in macrophages, lymphocytes and hepatocytes. Applications Reported: This 17D9 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 17D9 antibody has been tested by western blotting at 1-5 μg/mL on J774 cells or stimulated splenocyte lysates. Samples must be reduced for detection. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a protein that is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a prodomain and a large and small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. This caspase is able to cleave and activate its own precursor protein, as well as caspase 1 precursor. When overexpressed, this gene induces cell apoptosis. Alternative splicing results in transcript variants encoding distinct isoforms.
Specifications

Specifications

Caspase 11
0.5 mg/mL
Monoclonal
Liquid
RUO
P70343
IgG2a
Affinity Chromatography
Antibody
17D9
Western Blot
Unconjugated
Rat
Mouse
12363
Primary
4°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.