Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Caspase-12 Antibody, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
| Antigen | Caspase-12 | 
|---|---|
| Concentration | 0.5 mg/mL | 
| Dilution | Western Blot 0.25-1 ug/ml | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        NBP224518SS
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NBP224518SS  | 
                
            
            
            
            
            
                
                    0.025 mg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $181.90
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        NBP224518
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NBP224518  | 
                
            
            
            
            
            
                
                    0.1 mg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $402.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Caspase-12 Polyclonal specifically detects Caspase-12 in Mouse samples. It is validated for Western Blot.Specifications
| Caspase-12 | |
| Western Blot 0.25-1 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Mouse | |
| O08736 | |
| 100506742 | |
| Within the range of amino acids 100-131 of mouse Caspase-12 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | 
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415 | |
| CASP12 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in mouse and 94% homologous in rat. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title