Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-14 Antibody (001), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP305780PE
Description
Caspase-14 Monoclonal antibody specifically detects Caspase-14 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Caspase-14 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
001 | |
Immunohistochemistry-Paraffin | |
apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Caspase-14 (Accession#: NP_036246.1; Ser2-Gln242). | |
0.1 mL | |
Apoptosis, Cancer, Caspases | |
23581 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction