Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-14 Antibody (70A1426), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100567180.2ML
Description
Caspase-14 Monoclonal specifically detects Caspase-14 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Caspase-14 | |
Monoclonal | |
Unconjugated | |
Ascites with PBS with 0.09% Sodium Azide | |
apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
In mouse spleen, a 30 kDa band is observed. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry | |
70A1426 | |
Western Blot 0.1-4 ug/ml, Flow Cytometry (Intracellular): 1:500/10^6 cells | |
NP_033939 | |
CASP14 | |
This antibody was raised against a synthetic peptide corresponding to amino acids 2-18 of mouse Caspase-14. | |
0.2 mL | |
Apoptosis, Cancer, Caspases | |
23581 | |
Human, Mouse | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction