Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-14 Antibody, Novus Biologicals™
SDP

Catalog No. p-5053715 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056126T 0.025 mL
NB10056126 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056126T Supplier Novus Biologicals Supplier No. NB10056126SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Caspase-14 Polyclonal specifically detects Caspase-14 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Caspase-14
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Neat whole antisera with 0.02% Sodium Azide
Gene Accession No. NP_033939
Gene Alias apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE
Gene Symbols CASP14
Host Species Rabbit
Immunogen Recombinant full-length human Caspase-14 was used as immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 23581
Test Specificity This polyclonal antibody recognizes the proform of caspase-14 (∼-28-32 kDa), and the large (∼14-21 kDa) and small (∼10-11 kDa) of active/cleaved caspase-14.
Target Species Human, Mouse, Rat, Canine, Gerbil
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.