Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-14 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299890PE
Description
Caspase-14 Polyclonal antibody specifically detects Caspase-14 in Human samples. It is validated for ELISASpecifications
Caspase-14 | |
Polyclonal | |
ELISA | |
apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE | |
Produced in rabbits immunized with purified, recombinant Human Caspase-14 (Accession#: NP_036246.1; Ser2-Gln242) | |
0.1 mL | |
Apoptosis, Cancer, Caspases | |
23581 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction