Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 14 Monoclonal Antibody (05)
GREENER_CHOICE

Catalog No. PIMA529082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529082 Supplier Invitrogen™ Supplier No. MA529082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Caspase-14/CASP14.

Caspase 14 encodes a member of the cysteine-aspartic acid protease (caspase) family, which plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes that undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This caspase has been shown to be processed and activated by caspase 8 and caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in keratinocyte terminal differentiation, which is important for the formation of the skin barrier. Diseases associated with CASP14 include Ichthyosis, Congenital, Autosomal Recessive 12 and Autosomal Recessive Congenital Ichthyosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 14
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Casp14
Gene Accession No. P31944
Gene Alias apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE
Gene Symbols Casp14
Host Species Mouse
Immunogen Recombinant Human Caspase-14/CASP14 protein (Ser2-Gln242).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.