Learn More
Invitrogen™ Caspase 14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121009
Description
Positive test controls include: HepG2, 293T. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MSNPRSLEEE KYDMSGARLA LILCVTKARE GSEEDLDALE HMFRQLRFES TMKRDPTAEQ FQEELEKFQQ AIDSREDPVS CAFVVLMAHG REGFLKGEDG EMVKLENLFE ALNNKNCQAL RAKPKVYIIQ ACRGEQRDPG ETVGGDEIVM VIKDSPQTIP TYTDALHVYS TVEGYIAYRH DQKGSCFIQT LVDVFTKRKG HILELLTEVT RRMAEAELVQ EGKARKTNPE IQSTLRKRLY LQ.
Caspase 14 encodes a member of the cysteine-aspartic acid protease (caspase) family, which plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes that undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This caspase has been shown to be processed and activated by caspase 8 and caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in keratinocyte terminal differentiation, which is important for the formation of the skin barrier. Diseases associated with CASP14 include Ichthyosis, Congenital, Autosomal Recessive 12 and Autosomal Recessive Congenital Ichthyosis.
Specifications
| Caspase 14 | |
| Polyclonal | |
| Unconjugated | |
| Casp14 | |
| apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12365, 23581, 299587 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O89094, P31944 | |
| Casp14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-242 of human Caspase-14 (NP_036246.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.