Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 14 Recombinant Rabbit Monoclonal Antibody (145)
GREENER_CHOICE

Catalog No. PIMA529087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529087 Supplier Thermo Scientific Supplier No. MA529087
Add to Cart
Add to Cart

Caspase 14 Recombinant Rabbit Monoclonal Antibody (145)

Caspase 14 Recombinant Monoclonal Antibody for Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 14
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 145
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Casp14
Gene Accession No. P31944
Gene Alias apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE
Gene Symbols Casp14
Host Species Rabbit
Immunogen Recombinant Human Caspase-14/CASP14 protein (Ser2-Gln242).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.