Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-2 Antibody (10C6), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280095UV
This item is not returnable.
View return policy
Description
Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
Caspase-2 | |
Monoclonal | |
DyLight 350 | |
caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
Recombinant human caspase-2 (p19 subunit). | |
0.1 mL | |
Apoptosis, Cancer, Caspases | |
835 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
10C6 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction