Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-2 Antibody (18E809.3), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Caspase-2 |
---|---|
Clone | 18E809.3 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056686T
![]() |
Novus Biologicals
NB10056686SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056686
![]() |
Novus Biologicals
NB10056686 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
Caspase-2 Monoclonal specifically detects Caspase-2 in Human samples. It is validated for Western Blot.Specifications
Caspase-2 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
CASP2 | |
IgM κ | |
Unpurified | |
In HL60, a 48 kDa band is observed. This will cross-react with mouse and rat Caspase-2, based on BLAST. |
18E809.3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Caspases | |
P42575 | |
835 | |
A peptide corresponding to amino acids 400-450 of human Caspase-2 was used as immunogen. GenBank no NP_116764. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title