Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-2 Antibody (3D4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Caspase-2 |
---|---|
Clone | 3D4 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
Caspase-2 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
A synthesized peptide derived from Human Caspase-2 [UniProt P42575] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3D4 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Caspases | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
835 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title