Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Caspase 2 Monoclonal Antibody (18E809.3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA191729
Description
This antibody is expected to cross-react with mouse and rat Caspase-2 based on sequence homology.
Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Specifications
Caspase 2 | |
Monoclonal | |
Conc. Not Determined | |
Ascites, PBS with 0.2% gelatin and 0.05% sodium azide | |
P42575 | |
CASP2 | |
A peptide corresponding to amino acids 400-450 of human Caspase-2 was used as immunogen. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Western Blot | |
18E809 | |
Unconjugated | |
CASP2 | |
CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 | |
Mouse | |
IgM | |
RUO | |
835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction