Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-2 Rabbit anti-Human, Mouse, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF826SP
This item is not returnable.
View return policy
Description
Caspase-2 Polyclonal specifically detects Caspase-2 in Human, Mouse samples. It is validated for Western Blot.Specifications
Caspase-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CASP2 | |
KLH-coupled mouse Caspase-2 synthetic peptide KEREGYAPGTEFHRC | |
25 μg | |
Primary | |
Detects human and mouse Caspase-2 precursor and the small Caspase-2 subunit that is generated during proteolytic activation in Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.8 ug/mL | |
caspase-2, CASP2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, Caspase2, ICH1, ICH-1, NEDD2, NEDD-2, PPP1R57 | |
Rabbit | |
Affinity Purified | |
RUO | |
835 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction