Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Caspase 3/7 Apoptosis Kit
SDP

Catalog No. RD10188047 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10188047 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10188047 Supplier R&D Systems™ Supplier No. NBP350237

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Apoptosis Kit

Specifications

Quantity 100 Tests
Synonym CASP-3, CASP-7, caspase-3, caspase-7, CPP32, CPP32B, ICE-like apoptotic protease 3, SCA-1
Content And Storage Store at 4°C in the dark.
Product Line Novus
For Use With (Application) Detection Instrument: Flow Cytometer;Fluorescence Microscope Assay Time: 0.5 hour
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.