Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-3 Antibody (31A1067) - (Pro and Active) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233244
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Mammal samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.Specifications
Caspase-3 | |
Monoclonal | |
0.5mg/mL | |
Western Blot 1-5 ug/ml, Simple Western, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23840553), Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Lee et al), Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (Zhang et al), Knockout Validated, Knockdown Validated | |
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Mouse | |
31.7 kDa | |
0.1 mg | |
Apoptosis, Cancer, Caspases, Phospho Specific | |
836 | |
Human, Mouse, Rat, Pig, Chicken, Chinese Hamster, Mammalia | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
31A1067 | |
Unconjugated | |
P42574 | |
CASP3 | |
This Caspase-3 Antibody (31A1067) - (Pro and Active) - Azide and BSA Free was developed against full-length recombinant human caspase-3 protein. | |
Protein G purified | |
RUO | |
Primary | |
The antibody detects both pro Caspase-3 (∼32 kDa) and the large subunit of the active/cleaved form (∼14-21 kDa) of Caspase-3. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction