Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Antibody (31A1067) - (Pro and Active) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 155 publications

$247.53 - $591.43

Specifications

Antigen Caspase-3
Clone 31A1067
Concentration 1 mg/mL
Dilution Western Blot 1 - 5 ug/ml, Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 27429862), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23840553), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 28500555), Immunohistochemistry-Frozen 1:10 - 1:500. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 28500555), Hematoxylin and Eosin Stain reported in scientific literature (PMID 28186963), Electron Microscopy reported in scientific literature (PMID 27450722), Immunohistochemistry Free-Floating reported in scientific literature (PMID 31771656), CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 32867814)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Immunohistochemistry (Free Floating), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056708T
SDP
View Documents
Novus Biologicals
NB10056708SS
0.025 mg
Each for $247.53
Only null left
Add to Cart
 
NB10056708
SDP
View Documents
Novus Biologicals
NB10056708
0.1 mg
Each for $591.43
Only null left
Add to Cart
 
Description

Description

Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Mammal samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Hematoxylin & Eosin Stain, Electron Microscopy, Immunohistochemistry Free-Floating, CyTOF-ready, Knockout Validated, Knockdown Validated.
Specifications

Specifications

Caspase-3
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Immunohistochemistry (Free Floating), CyTOF
Unconjugated
Mouse
Apoptosis, Cancer, Caspases, Phospho Specific
P42574
836
This Caspase-3 Antibody (31A1067) - (Pro and Active) was developed against full-length recombinant human caspase-3 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
31.7 kDa
31A1067
Western Blot 1 - 5 ug/ml, Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 27429862), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23840553), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 28500555), Immunohistochemistry-Frozen 1:10 - 1:500. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 28500555), Hematoxylin and Eosin Stain reported in scientific literature (PMID 28186963), Electron Microscopy reported in scientific literature (PMID 27450722), Immunohistochemistry Free-Floating reported in scientific literature (PMID 31771656), CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 32867814)
Monoclonal
Purified
RUO
Human, Mouse, Rat, Pig, Chinese Hamster, Mammalia
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
CASP3
IgG1 κ
Protein G purified
The antibody detects both pro Caspase-3 (∼32 kDa) and the large subunit of the active/cleaved form (∼14-21 kDa) of Caspase-3.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.