Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Antibody - (active/cleaved), Novus Biologicals™
SDP

Catalog No. p-5054290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056113T 0.025 mL
NB10056113 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056113T Supplier Novus Biologicals Supplier No. NB10056113SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 48 publications

Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).

Specifications

Antigen Caspase-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry reported in scientific literature (PMID 28287161), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29963272), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen reported in scientific literature (PMID 29864441), Immunoblotting reported in scientific literature (PMID 27912032), Flow (Intracellular) reported in scientific literature (PMID 24804954)
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Alias Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Gene Symbols CASP3
Host Species Rabbit
Immunogen This Caspase-3 Antibody - (active/cleaved) was developed against catalytically active human caspase-3 protein.
Molecular Weight of Antigen 31.7 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 836
Test Specificity Preferentially detects active caspase-3 (large subunit: ∼14-21 kDa, and small subunit: ∼10 kDa). However, it may also detect pro-caspase-3 (∼32 kDa) in some cell or tissue systems. Nuclear immunostaining of caspase-3 is considered be an indication of active/cleaved caspase-3.
Target Species Human, Mouse, Rat, Gerbil
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.