Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB6001235
Description
Caspase-3 Polyclonal antibody specifically detects Caspase-3 in Human, Fish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating.Specifications
Caspase-3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Rabbit | |
31.7 kDa | |
0.5 mL | |
Primary | |
This reacts with the active form of Caspase 3 (17 kDa protein). | |
Store at 4C. Do not freeze. | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 | |
P42574, P42574 | |
CASP3 | |
This Caspase-3 Antibody was developed against a synthetic peptide corresponding to the cleavage site of human caspase 3 (amino acids 167-175), conjugated to KLH. | |
Affinity purified | |
RUO | |
836 | |
Human, Fish, Rat, Pig, Bovine, Canine, Fish, Monkey, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction