Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Antibody - (Pro and Active), Novus Biologicals™
SDP

Catalog No. NB10056112T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056112T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056112T Supplier Novus Biologicals Supplier No. NB10056112SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 14 publications

Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Caspase-3
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Simple Western 1:200, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 27735949), Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P42574
Gene Alias Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Gene Symbols CASP3
Host Species Rabbit
Immunogen This Caspase-3 Antibody - (Pro and Active) was developed against full-length human Caspase-3 protein (pro-form).
Molecular Weight of Antigen 31.7 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 836
Test Specificity Caspase antibodies are classical tools for detecting inactive (pro) and active (cleaved) forms of the enzymes. The presence of the large or small subunits in western blots is considered to be a marker of caspase activation.
Target Species Human, Mouse, Rat, Canine, Gerbil
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.