Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 3 Monoclonal Antibody (MAR-1), eBioscience™
GREENER_CHOICE

Catalog No. 5017709
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-177-09 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-177-09 Supplier Invitrogen™ Supplier No. BMS1040
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: Caspases are present in all cells, recruitment of these proteases to oligomerized receptors leads to activation accompanied by autoproteolytic cleavage. Active caspases can proteolyze additional caspases generating a caspase cascade that cleaves proteins critical for cell survival. The final outcome of this signaling pathway is a form of controlled cell death termed apoptosis. The subgroup of caspases involved in apoptosis is called initiators or effectors. Caspase-3 cleaves substrate at the carboxyl terminus of aspartate residues. Active caspase-3 has two active sites and consists of two identical large (~ 20 kDa) and two identical small (~ 10 kDa) subunits that are derived from two precursor caspase-3 polypeptides. Caspase-3 is proteolytically activated by other caspases. Both subunits contribute to substrate binding and catalysis. The active site cysteine that covalently binds the substrate is located near the C-terminus of the large subunit. Active caspase-3 has two-fold symmetry, two active site pockets each residing on an opposite side. Caspase-3, together with caspases 8 and 9, is situated at pivotal junctions in apoptotic pathways. Caspase-3 appears to amplify caspase 8 and 9 initiation signals into complete commitment to apoptotic disassembly. Applications Tested: ELISA, Western Blotting.

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 6, 7 and 9, and the protein itself is processed by caspases 8, 9 and 10. It is the predominant caspase involved in the cleavage of amyloid-beta 4A precursor protein, which is associated with neuronal death in Alzheimer's disease. Alternative splicing of this gene results in two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 3
Applications ELISA, Western Blot
Classification Monoclonal
Clone MAR-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2-7.4
Gene CASP3
Gene Accession No. P42574
Gene Alias A830040C14Rik; AC-3; Apopain; Apopain precursor; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase3; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; Caspase-3-like protein; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; cysteine protease; CPP32; apopain; ICE3; IRP; LICE; mldy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein
Gene Symbols CASP3
Host Species Mouse
Immunogen Rec. Human Caspase-3.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 836
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.