Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 3 p17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578921
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578921 Supplier Invitrogen™ Supplier No. PA578921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Heart Tissue, Rat Liver Tissue, Rat Thymus Tissue, Rat Spleen Tissue, mouse thymus tissue, mouse spleen tissue, mouse lung tissue, mouse brain tissue, mouse testis tissue. IHC: rat intestine tissue.

CASP3 (Caspase3 Apoptosis-Related Cysteine Protease), also known as YAMA, CPP32 or APOPAIN, is a caspase protein that interacts with caspase 8 and caspase 9. It is encoded by the CASP3 gene. The CASP3 protein is a member of the cysteine-aspartic acid protease (caspase) family. Tiso et al. (1996) used radiation hybrid mapping to localize the CPP32 gene to human chromosome 4q33-q35.1. Fernandes-Alnemri et al. (1994) found that overexpression of CPP32 in insect cells induced apoptosis. Coexpression of the 2 CPP32 subunits in insect cells also resulted in apoptosis. Tewari et al. (1995) showed that purified human ICE cleaved the Yama proenzyme into a proteolytically active form and that activated Yama cleaved PARP into the 85-kD apoptotic form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 3 p17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene CASP3
Gene Accession No. P55213, P70677
Gene Alias A830040C14Rik; AC-3; Apopain; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CC3; Cleaved Caspase 3; Cpp32; CPP-32; CPP32beta; cysteine protease CPP32; IRP; LICE; mldy; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein
Gene Symbols CASP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of mouse Caspase-3 (P17) (67-81aa TDVDAANLRETFMGL).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12367, 25402
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.