Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-3 Rabbit anti-Human, Mouse, Rat, Clone: 1U1D9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Caspase-3 |
---|---|
Clone | 1U1D9 |
Dilution | Western Blot 1:500 - 1:1000, Knockout Validated |
Applications | Western Blot, KnockDown |
Classification | Monoclonal |
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
Caspase-3 | |
Western Blot 1:500 - 1:1000, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Recombinant fusion protein containing a sequence corresponding to amino acids 29-175 of human Caspase-3 (P42574). SGISLDNSYKMDYPEMGLCIIINNKNFHKSTGMTSRSGTDVDAANLRETFRNLKYEVRNKNDLTREEIVELMRDVSKEDHSKRSSFVCVLLSHGEEGIIFGTNGPVDLKKITNFFRGDRCRSLTGKPKLFIIQACRGTELDCGIETD | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1U1D9 | |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
836 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title