Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-4 Antibody (OTI4A2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270339AF350
Description
Caspase-4 Monoclonal specifically detects Caspase-4 in Human samples. It is validated for Western Blot.Specifications
Caspase-4 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CASP4 | |
Recombinant protein fragment corresponding to amino acids 81-270 of human Caspase-4 (NP_150649) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI4A2 | |
Western Blot | |
apoptotic cysteine protease Mih1/TX, CASP-4, caspase 4, apoptosis-related cysteine peptidase, caspase 4, apoptosis-related cysteine protease, caspase-4, EC 3.4.22, EC 3.4.22.57, ICE(rel)II, ICE(rel)-II, ICEREL-II, ICH2, ICH-2, Mih1/TX, Protease ICH-2, Protease TX, TX | |
Mouse | |
0.1 mL | |
837 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction