Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518300 Supplier Invitrogen™ Supplier No. PA518300
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, feline and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a protein that is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a prodomain and a large and small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. This caspase is able to cleave and activate its own precursor protein, as well as caspase 1 precursor. When overexpressed, this gene induces cell apoptosis. Alternative splicing results in transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CASP4
Gene Accession No. P49662
Gene Alias apoptotic cysteine protease Mih1/TX; Casp11; CASP-11; CASP4; CASP-4; caspase 11; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-11; Caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspl; ICE and Ced-3 homolog 2; ICE(rel)II; ICE(rel)-II; ICEREL-II; ICH2; ICH-2; Ich3; ich-3; Mih1; Mih1/TX; protease ICH-2; Protease ICH-3; Protease TX; TX
Gene Symbols CASP4
Host Species Goat
Immunogen Synthetic peptide sequence (CKERAEEIYPIKERN) corresponding to the internal amino acids of CASP4 (aa 118-133).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 837
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.