Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520110 Supplier Invitrogen™ Supplier No. PA520110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Ramos cell lysate. PA5-20110 can be used with blocking peptide PEP-0228.

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12. Activation of these caspases results in the cleavage and activation of proinflammatory cytokines such as IL-1-b and IL-18. Caspase-5 can interact with caspase-1; both are constituents of the NALP1 inflammasome, a complex that can trigger the cleavage of pro-IL-1-b. Expression of caspase-5 can be regulated by lipopolysaccharide (LPS) and IFN-gamma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CASP5
Gene Accession No. P51878
Gene Alias CASP5; CASP-5; caspase 5; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE(rel)III; ICE(rel)-III; ICEREL-III; ICH3; ICH-3; Protease ICH-3; protease TY; TY protease
Gene Symbols CASP5
Host Species Rabbit
Immunogen A 16 amino acid peptide from the amino-terminus of human Caspase-5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 838
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.