Specifications
| Quantity | 100 Tests |
| Content And Storage | 100 mL Cell Lysis Buffer, 4 x 2 mL 2X Reaction Buffer, 0.5 mL WEHD-AFC (1 mM), & 0.4 mL DTT (1 M). Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |