Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 6 Monoclonal Antibody (C1)
GREENER_CHOICE

Catalog No. PIMA541562
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541562 200 μL
PIMA543919 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541562 Supplier Invitrogen™ Supplier No. MA541562
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caspase 6 Monoclonal Antibody for Western Blot, IHC (P)

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 6
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Casp6
Gene Accession No. Q3T0P5
Gene Alias apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2
Gene Symbols Casp6
Host Species Mouse
Immunogen Recombinant protein Caspase 6.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 538409
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.