Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 6 Monoclonal Antibody (M378)
GREENER_CHOICE

Catalog No. PIMA547700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547700 Supplier Invitrogen™ Supplier No. MA547700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 34 kDa* protein corresponding to the molecular weight of Caspase-6 on SDS-PAGE immunoblots of human Jurkat, K562, and A431 cells.

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 6
Applications Western Blot
Classification Monoclonal
Clone M378
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Casp6
Gene Accession No. P55212
Gene Alias apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2
Gene Symbols Casp6
Host Species Mouse
Immunogen Clone (M378) was generated from a peptide containing amino acid residues in the C-terminal region of human Caspase-6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 839
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.