Learn More
Invitrogen™ Caspase 6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143078
Description
Additional Information: This antibody is expected to recognize reproted isoform alpha (NP_001217.2) only. The immunizing peptide overlaps phosphorylated serine 257 and therefore this antibody is not expected to recognize the phosphorylated protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Specifications
| Caspase 6 | |
| Polyclonal | |
| Unconjugated | |
| Casp6 | |
| apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 839 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55212 | |
| Casp6 | |
| Peptide with sequence NRKVSQRRVDFCKDP (aa 253-267). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.