Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-7 Antibody (7-1-11), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP119229AF488
Description
Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat, Porcine, Canine, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Caspase-7 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CASP7 | |
Recombinant human Caspase-7 protein (full length) | |
0.1 mL | |
Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway | |
Caspase-7 (7-1-11) | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
7-1-11 | |
Western Blot, Immunohistochemistry | |
apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Hamster, Primate, Rabbit, Sheep | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction